Learn More
Greiner Bio-One 96-well Chimney designed Round (U) Bottom Microplates
Catalog No. 07000146 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
Blue
Clear
Green
Natural
Red
White
Yellow
Sterility:
Non-sterile
Sterile
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility
07-000-146 Clear Non-sterile
07-000-143 Black Non-sterile
07-000-150 Natural Sterile
07-000-996 Clear Sterile
07-000-144 Blue Non-sterile
07-000-145 Green Non-sterile
07-000-147 Red Non-sterile
07-000-149 Yellow Non-sterile
07-000-148 White Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 07-000-146 Supplier Greiner Bio-One Supplier No. 650201
Add to Cart
Add to Cart
For high-throughput screening in cell based test systems.
  • Non-pyrogenic
  • Conical well design with flat bottom
  • Ensures accurate pipetting
  • Reduces dead volume of well
  • Free of detectable DNase, RNase, human DNA
  • Prevents wicking
TRUSTED_SUSTAINABILITY
Specifications
Breakapart No
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Binding Type Non Binding
Color Clear
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 355 μL
Well Shape Round Bottom (U-well)
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 100
Quantity 100/Cs.
Well Color Clear
Well Design Chimney