Learn More
Thermo Scientific™ 96-Well Microtiter™ Microplates
Catalog No. 14245142
Change view
Click to view available options
Material:
Polystyrene
Vinyl
Well Shape:
Conical Bottom (V-well)
Curved Edges, Flat Bottom (C-well)
Flat Bottom (F-well)
Round Bottom (U-well)
Sterility:
Non-sterile
Sterile
Volume (Metric) Well:
280 μL
330 μL
400 μL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Well Shape Sterility Volume (Metric) Well
14-245-142 Vinyl Flat Bottom (F-well) Non-sterile 330 μL
14-245-71 Polystyrene Round Bottom (U-well) Non-sterile 400 μL
14-245-72 Polystyrene Conical Bottom (V-well) Non-sterile 280 μL
14-245-145 Polystyrene Curved Edges, Flat Bottom (C-well) Non-sterile 330 μL
14-245-140 Vinyl Round Bottom (U-well) Non-sterile 400 μL
14-245-73 Vinyl Round Bottom (U-well) Sterile 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-245-142 Supplier Thermo Scientific™ Supplier No. 2801
Use these 96-well microplates for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
Thermo Scientific™ 96-Well Microtiter Microplates are ideal for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
  • For high-throughput screening and research
  • Clear plates for colorimetric applications
  • Standard footprint, 128 x 86 mm
  • Available as both polystyrene square wells and vinyl square wells
  • Vinyl Microtiter microplates are used for complement fixation, agglutination, and blood typing
  • Styrene Microtiter microplates are used for complement fixation, agglutination, microbiology testing, and blood typing
Ordering Information:
Polystyrene microplates are available as 50 units/package. Vinyl microplates are available as 25 or 100 units/package, depending on product.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Lid Without Lid
Array 8 x 12
Packaging 100 Pk.
Product Line Microtiter
Color Clear
Product Type Microplate
Material Vinyl
Volume (Metric) Well 330 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
Quantity 100/Pk.