Learn More
Greiner Bio-One 96-well Non-treated Polypropylene Microplates
GREENER_CHOICE
Catalog No. 07000110 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
Clear
Green
Pink
Sterility:
Non-sterile
Sterile
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility Well Shape
07-000-110 Black Non-sterile Flat Bottom (F-well)
07-000-153 Clear Non-sterile Conical Bottom (V-well)
07-000-113 Clear Non-sterile Flat Bottom (F-well)
07-000-117 Clear Sterile Flat Bottom (F-well)
07-000-114 Pink Non-sterile Flat Bottom (F-well)
07-000-112 Green Non-sterile Flat Bottom (F-well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 07-000-110 Supplier Greiner Bio-One Supplier No. 655209
Add to Cart
Add to Cart
This item is not returnable. View return policy
Features high chemical resistance. Greiner Bio-One 96-Well Non-treated Polypropylene Microplates is free of detectable DNase, Rnase and human DNA.
  • Non-Pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Material: Polypropylene
  • High chemical resistance
  • Sealable with adhesive films and heat sealer
  • Can be traced back to production through defined LOT nu
Certifications
DIN ISO 9001
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Color Black
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
Quantity 100/Cs.
Well Design Chimney