Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Thermo Scientific™ 96-well Pierceable Seals

Catalog No. A55331 Shop All Thermo Scientific Products
Change view
Click to view available options
Material:
Polyester
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
A55331 Polyester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A55331 Supplier Thermo Scientific™ Supplier No. A55331
Add to Cart
Add to Cart
This item is not returnable. View return policy

Eliminate lysis tube capping and uncapping steps with the specifically designed Thermo Scientific 96-well Pierceable Seals in the SureTect workflow.

Speed up the lysis and pipetting steps during the SureTect™ workflow using the Thermo Scientific™ 96-well Pierceable Seals.

Designed specifically for use with the SureTect™ assays the 96-well Pierceable Seals offer the ability to quickly and securely seal and pipette from the lysis tubes, reducing cross contamination and eliminating the requirement for capping and uncapping steps to enable a simple, streamlined workflow for maximum efficiency and productivity in your laboratory.

Use the Thermo Scientific 96-well Pierceable Seals to streamline your SureTect PCR Assay workflow, quickly and easily sealing lysis plates and pipette from the SureTect lysis tubes directly through the seal, reducing cross-contamination and eliminating the need for capping and uncapping to enable a fast and easy to use workflow.

The high integrity sealing materials feature a strong adhesive providing secure plate sealing during cycling and sample storage.

Printed well positions and orientation markings enable you to position and identify samples

Strip perforations for ease of separation and use regardless of your sample throughput.

Combined with our comprehensive portfolio of assays and world-class instrumentation you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.

We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Thermo Scientific™ SureTect™ Real-Time PCR System is continually evolving to support your laboratory and testing needs.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Description 96-well Pierceable Seals
Material Polyester
For Use With (Application) SureTect PCR Assay workflow
Pierceable Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.