Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accumax Lab Devices 96 Well Plate Optical Sealing Membrane

Catalog No. 02945149
Change view
Click to view available options
For Use With (Application):
PCR
qPCR
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
02-945-149 PCR
02-945-148 qPCR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-945-149 Supplier Accumax Lab Devices Supplier No. 335815
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These consumables are highly heat resistant up to 121°C thus making it compatible with every type of PCR reaction.

  • Strong adhesive, prevents evaporation during thermal cycles
  • Temperature stability range from -20 to 120°C
  • Highly transparent and clear for sample inspection
  • Doesn't interfere with the luminescence and fluorescence assay
  • Non pierceable, better protection against accidental piercing
  • Easy adhesive film, no pressure required while applying

Specifications

Color Clear
Sterility Sterile
Autoclavable Autoclavable
No. per Case 100
No. per Pack 10
For Use With (Application) PCR
Product Type Well Plate Seal
Adhesive Adhesive Clear Seal
Pierceable No
Fit for Purpose PCR Ready
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.