Learn More
Applied Biosystems™ Plate Septa, 96 well
Description
Includes
Each package contains 20 septa, sufficient for 1,920 samples
These are septa for use with the 3730, 3730xl, 3130xl, and 3130, instruments.
Use with:
310 Running Buffer (10X), 3100/3100-Avant™ Genetic Analyzer Autosampler Plate Kit, 96-well, 3130/3100-Avant™ Genetic Analyzer Capillary Array (36 cm), 3130xl/3100 Genetic Analyzer Capillary Array (36 cm), 3130xl/3100 Genetic Analyzer Capillary Array (50 cm), 3730 Running Buffer (10X), 5.0 mL Glass Syringe (polymer reserve syringe), 96-Well Plate Base, 96-Well Plate Retainer, Hi-Di™ Formamide, MicroAmp™ Optical 96-Well Reaction Plate, MicroAmp™ Optical 96-Well Reaction Plate with Barcode, POP-4™ Polymer for 3100/3100-Avant™ Genetic Analyzers, POP-4™ Polymer for 3130/3130xl Genetic Analyzers, POP-4™ Polymer for 3130/3130xl Genetic Analyzers, POP-7™ Polymer for 3130/3130xl Genetic Analyzers, POP-7™ Polymer for 3130/3130xl Genetic Analyzers, Reservoir Septa
Specifications
Specifications
| Quantity | 20 each |
| Format | 96-well Plate |
| For Use With (Equipment) | 3130xl Genetic Analyzer, 3730xl DNA Analyzer, 3730 DNA Analyzer, 3130 Genetic Analyzer |
| Product Type | Septa Plate |
| Shipping Condition | Room Temperature |
| Content And Storage | Each package contains 20 septa, sufficient for 1,920 samples. |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.