Learn More
Thomas Scientific Pre-Slit Pierceable Film, 100/pk
SDP
Supplier:  Thomas Scientific 1150Q09
Encompass
Ideal for short-term storage at ambient temperatures and robotics. Designed for short-term handling and storage of samples that require multiple access to wells by pipet tips or robotic probes. A pre-cut “X” over each well creates bendable flaps that provide sample access without coring or adhesive fouling of tips or probes.
Catalog No. NC1718807