Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ 96-Well Protein Precipitation Filter Plate
Ideal for removing precipitated proteins from biological plasma/serum
Supplier: MilliporeSigma™ Supelco™ 55263U
Description
- Plate consists of a 0.2 μm hydrophobic graded filter/frit
- Biological plasma is first added to the 96-well plate followed by a protein precipitating agent (e.g., acetonitrile)
- After a brief mixing step, vacuum is applied to the plate, and the filter/frit removes precipitated proteins from the sample
- The resulting filtrate is ready for further processing and/or analysis
Specifications
| Hydrophobic Graded Filter/Frit | |
| Protein Precipitation Filter Plate | |
| 96 Wells | |
| 1/Pk. |
| Removing Precipitated Proteins From Biological Plasma/Serum | |
| 2 mL | |
| 0.2 μm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction