Learn More
Greiner Bio-One 96-well Sapphire Microplates
Catalog No. 07000312 Shop All Greiner Bio-One Products
Change view
Click to view available options
Skirt Style:
Full
Half
Non-skirt
No. per Case:
40
100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Skirt Style No. per Case
07-000-312 Non-skirt 100
07-000-105 Full 100
07-000-313 Half 100
07-000-107 Half 40
07-000-106 Non-skirt 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 07-000-312 Supplier Greiner Bio-One Supplier No. 652250
Add to Cart
Add to Cart
Features optimal heat transfer from thermoblock to reaction solution.
  • >Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Ultra thin polypropylene for optimal heat transfer
  • Are particularly suited for automation
  • Robot-friendly- for grippers, sufficient space for the application
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Packaging Inner bags of 10
Color Clear
Skirt Style Non-skirt
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
No. per Case 100
Plate Compatibility Universal
Quantity 100/Cs.