Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ 96-Well Semi-Skirted PCR Plates

Catalog No. 14230246 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-230-246 Blue
14-230-248 Yellow
14-230-245 Red
14-230-244 Natural
14-230-247 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-230-246
Add to Cart
Add to Cart

Directly compatible with all major thermal cyclers, including ABI instruments—can be used in ABI 96-well sequencers without adapters

  • Virgin polypropylene
  • Clear well bottom for sample visibility
  • Raised-rim design
  • Sealable with adhesive films and foils, heat seals, and cap strips
  • 300μL maximum volume when used with adhesive and heat seals
  • Cut-away corner and alphanumeric grid
  • Cleanroom produced

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Array 8 x 12
Color Blue
Product Type PCR Microplate
Skirt Style Semi
Material Polypropylene
Volume (Metric) Well 300 μL
No. of Wells 96
For Use With (Application) PCR, RT-PCR applications
Rimmed Yes
Well Color Clear
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.