Learn More
Cytiva 96 well Septa
Catalog No. 501528208 Shop All Cytiva Products
Encompass
Septa to cover 96-well microplates used in Biacore 8k, Biacore T200 and Biacore S200.
Catalog No. 50-152-8208 Supplier Cytiva Supplier No. 29192561
Septa to cover 96-well microplates used in Biacore 8k, Biacore T200 and Biacore S200.