Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Septa for 96-Well Plates, for 3500/SeqStudio™ Flex

Catalog No. 4412614
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-126-14 20 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-126-14 Supplier Applied Biosystems™ Supplier No. 4412614
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

These septa for 96 well plates are designed and validated for use with Applied Biosystems 3500 and SeqStudio Flex Series Genetic Analyzers.

These septa for 96 well plates are designed and validated for use with Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 20 each
Format 96-well Plate
For Use With (Equipment) 3500 Genetic Analyzer, 3500xL Genetic Analyzer
Product Type Septa Plate
Shipping Condition Room Temperature
Content And Storage One bag of 20 septa. Store at room temperature.
How often can the plate septa on the 3130/xl, 3730/xl, 3500/xl, and SeqStudio instruments be used?

We officially recommend that the plate septa be used one time only. However, we are aware that some of our customers will rinse the septa with water and re-use them. In those instances, the septa can become sticky and gummy over time and eventually start sticking to the array. In the best case scenario, the array will lift the plate off of the autosampler just enough to break the connection with the plate sensor and abort the run. The Event Log might show that the run aborted due to the user aborting it, so the run will appear to have completed or stopped with no errors and no data. The worst case scenario is that the plate remains on the array as the autosampler starts to move, bending the electrodes and breaking the array. We recommend replacing the plate septa regularly (replacing after each run is recommended). Do not use bleach, detergents, or solvents to clean the septa, it will cause them to dry out faster and also risks contaminating the system.

When I started my run on the Applied Biosystems 3500/3500xL Genetic Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?

Some of the other causes of an Unstable Electrophoresis current detected error message are:

1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues

Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.

Do I need to replace the buffer every 14 days if the Applied Biosystems 3500/3500xL Genetic Analyzer is not in use?

If the instrument is not in use, it is not necessary to replace the buffer every 14 days. However, the buffer level needs to remain at the fill line to prevent the capillary or array from drying out. Top off the buffer volume with water if necessary. Replace the anode and cathode buffer containers prior to starting a new run.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.