Learn More
Greiner Bio-One 96-well Solid Bottomed Microplates
Catalog No. 07000132 Shop All Greiner Bio-One Products
Change view
Click to view available options
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round Bottom (U-well)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape
07-000-132 Round Bottom (U-well)
07-000-125 Flat Bottom (F-well)
07-000-142 Conical Bottom (V-well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-000-132 Supplier Greiner Bio-One Supplier No. 650161
Add to Cart
Add to Cart
For high-throughput screening and assay
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Ideally suited for agglutination tests
  • No sharp corners, which means that they can be pipetted easily and cleanly
  • Suitable for +/- analyses
  • Solid bot
TRUSTED_SUSTAINABILITY
Specifications
Breakapart No
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type Non Binding
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 40 to 280 μL
Sterility Sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 323 μL
Well Shape Round Bottom (U-well)
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 100
Quantity 100/Cs.
Well Color Clear
Well Design Chimney