Learn More
Kuhner Shaker Inc 96 WELL SQUARE GLUCOSE FEED
Supplier:  Kuhner Shaker Inc SMFP08002
Encompass_Preferred
FeedPlate, Glucose, 96 Well, Square, 14 mg/48h - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!
Catalog No. NC2478452
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.