Learn More
Corning™ 96-well, Ultra-low Binding, U-shaped Bottom Microplate PROMO
Catalog No. 12456721S24 Shop All Corning Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
12-456-721S24 Black
1 options
This item is not returnable. View return policy
Catalog No. 12-456-721S24 Supplier Corning™ Supplier No. 12456721S24
This item is not returnable. View return policy
With their novel and proprietary design, these 96-well, Ultra-low Binding Microplates are ideal for generating and analyzing 3D multicellular spheroids in the same microplate.
  • Ultra-Low Attachment (ULA) surface enables uniform and reproducible 3D multicellular spheroid formation
  • Black opaque microplate body shields each optically clear, round bottom well from well-to-well cross-talk
  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
  • Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization
  • You can culture and assay spheroids in the same microplate, without the need for transfer to a new microplate
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid With Lid
Array 8 x 12
Binding Type Ultra-low Attachment
Color Black
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 0.075 to 0.3 mL
Sterility Sterile
Surface Treatment Ultra-low Binding
Total Volume (Metric) Well 300 μL
Well Shape Round Bottom (U-well)
Includes Plate with Closure
No. per Case 5 Plates/Cs.
No. per Pack 1 Plate/Pk.