Learn More
Thomas Scientific 96 Well 1.1mL Deep Well Plate, Round Bottom, 50 case
SDP
Supplier:  Thomas Scientific 1149J82
Encompass
Ideal for sample collection, preparation, and long-term storage. HTS-friendly—compatible with most liquid-handling robots. Centrifugation to 4,000 x G. Manufactured from virgin polypropylene to ensure purity and sample integrity, chemical resistance, and compatibility with long-term storage needs.
Catalog No. NC1757610
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.