Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ammonium nickel(II) sulfate hexahydrate

Catalog No. A1827500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1827500G 500 g
A1827100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1827500G Supplier Sigma Aldrich Supplier No. A1827500G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ammonium nickel(II) sulfate hexahydrate

Specifications

Melting Point 85°C to 89°C
CAS 7785-20-8
Quantity 500 g
Molecular Formula H8N2NiO8S2 -+ 6H2O
Linear Formula (NH4)2 Ni(SO4)2 -+ 6H2O
MDL Number MFCD00150260
Synonym Nickel ammonium sulfate
Molecular Weight (g/mol) 394.99
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.