Learn More
LIFE TECHNOLOGIES TRIZOL TUBES 50ML
Catalog No. A33254
Encompass_Preferred
Catalog No. A33254 Supplier LIFE TECHNOLOGIES Supplier No. A33254
Can Phasemaker Tubes be used for isolation of DNA or protein?
No. Phasemaker tubes are only validated for RNA extraction.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov