Learn More
Description
- 4-(2-aminoethyl)morpholine is widely used in biomedical applications as this moiety serves as an important lysosome-targeting group. some of its applications include:
- Synthesis of the lysosome-targetable fluorescent probe for hydrogen sulfide imaging in living cells
- Synthesis of 1,8-naphthalimide conjugated troger's bases as deoxyribonucleic acid (DNA) targeting fluorescent probe
- Synthesis of intramolecular charge transfer-photoinduced electron transfer-fluorescence resonance energy transfer (ict-pet-fret) fluorescent probe for monitoring pH changes in living cells
- It is also used as a precursor to synthesize a variety of antimicrobial agents
Specifications
Specifications
| Density | 0.992 g/mL (at 25°C (literature)) |
| Boiling Point | 205°C (lit.) |
| Molecular Formula | C6H14N2O |
| Refractive Index | n20/D 1.476 (literature) |
| Linear Formula | C6H14N2O |
| MDL Number | MFCD00006182 |
| Quantity | 25 g |
| Synonym | 2-Morpholinoethylamine |
| Molecular Weight (g/mol) | 130.19 |
| Percent Purity | 99% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.