Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-chlorobutyrate, 98%

Catalog No. AAA1107036
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1107036 500 g
AAA1107022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1107036 Supplier Thermo Scientific Chemicals Supplier No. A1107036
Add to Cart

CAS: 3153-36-4 | C6H11ClO2 | 150.602 g/mol

Ethyl 4-chlorobutyrate is used in organic synthesis. And also used for producting 4-Iodo-butyric acid ethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-chlorobutyrate is used in organic synthesis. And also used for producting 4-Iodo-butyric acid ethyl ester.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3153-36-4
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.602
MDL Number MFCD00001004
InChI Key OPXNFHAILOHHFO-UHFFFAOYSA-N
Synonym ethyl 4-chlorobutyrate, butanoic acid, 4-chloro-, ethyl ester, ethyl gamma-chlorobutyrate, ethyl-4-chlorobutyrate, 4-chloro-butyric acid ethyl ester, butyric acid, 4-chloro-, ethyl ester, ethyl .gamma.-chlorobutyrate, 4-chlorobutyric acid ethyl ester, ethyl 4chlorobutyrate, acmc-1cqjw
PubChem CID 18474
IUPAC Name ethyl 4-chlorobutanoate
SMILES CCOC(=O)CCCCl

Specifications

Density 1.077
Boiling Point 185°C to 186°C
Flash Point 76°C (169°F)
Refractive Index 1.432
Quantity 500 g
Beilstein 1098810
Solubility Information Insoluble in water.
Formula Weight 150.61
Percent Purity 98%
Chemical Name or Material Ethyl 4-chlorobutyrate
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 221-591-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.