Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorophenylacetic acid, 98+%

Catalog No. AAA1121006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1121006 5 g
AAA1121014 25 g
AAA1121022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1121006 Supplier Thermo Scientific Chemicals Supplier No. A1121006
Add to Cart

CAS: 1878-65-5 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-65-5
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00004332
InChI Key WFPMUFXQDKMVCO-UHFFFAOYSA-N
Synonym 3-chlorophenylacetic acid, m-chlorophenylacetic acid, 2-3-chlorophenyl acetic acid, 3-chlorobenzeneacetic acid, benzeneacetic acid, 3-chloro, m-chlorophenyl acetic acid, 3-chlorophenyl acetic acid, 3-chlorophenylaceticacid, acetic acid, m-chlorophenyl
PubChem CID 15879
IUPAC Name 2-(3-chlorophenyl)acetic acid
SMILES OC(=O)CC1=CC=CC(Cl)=C1

Specifications

Melting Point 77°C to 80°C
Quantity 5 g
Odor Odorless
Beilstein 2045103
Formula Weight 170.6
Percent Purity ≥98%
Chemical Name or Material 3-Chlorophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-520-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.