Learn More
Trimethylene oxide, 97%
Description
Trimethylene oxide useful for the synthesis of 3-substituted propanols by reaction with Grignards or organolithiums in the presence of CuBr.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Trimethylene oxide useful for the synthesis of 3-substituted propanols by reaction with Grignards or organolithiums in the presence of CuBr.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is hygroscopic in nature and sensitive to air. Store under inert gas.
Specifications
Specifications
| Melting Point | -97°C |
| Density | 0.891 |
| Boiling Point | 49°C to 50°C |
| Flash Point | −28°C (−18°F) |
| Refractive Index | 1.393 |
| Quantity | 25 g |
| UN Number | UN1993 |
| Beilstein | 102382 |
| Merck Index | 14,9715 |
| Solubility Information | Soluble in water. |
| Show More |
Safety and Handling
| Hazard Category | H225-H302+H312+H332 |
| Hazard Statement | GHS H Statement H225-H302-H312-H332 Highly flammable liquid and vapour. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 207-964-3 |
| RTECSNumber | RQ6825000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.