Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[h]quinoline, 98%

Catalog No. AAA1779403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1779403 1 g
AAA1779406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1779403 Supplier Thermo Scientific Chemicals Supplier No. A1779403
Add to Cart

CAS: 230-27-3 | C13H9N | 179.222 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 230-27-3
Molecular Formula C13H9N
Molecular Weight (g/mol) 179.222
MDL Number MFCD00004984
InChI Key WZJYKHNJTSNBHV-UHFFFAOYSA-N
Synonym benzo h quinoline, 7,8-benzoquinoline, 4-azaphenanthrene, 1-naphthoquinoline, benzoquinoline, alpha-naphthoquinoline, alpha-benzoquinoline, 7,8-bnzoquinoline, .alpha.-benzoquinoline
PubChem CID 9191
IUPAC Name benzo[h]quinoline
SMILES C1=CC=C2C(=C1)C=CC3=C2N=CC=C3

Specifications

Melting Point 48°C to 52°C
Boiling Point 233°C (47 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 120249
Formula Weight 179.22
Percent Purity 98%
Chemical Name or Material Benzo[h]quinoline
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 205-937-0
RTECSNumber DK1432000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.