Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-2,4,6-triiodobenzoic acid, 97%

Catalog No. AAB2335522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2335522 100 g
AAB2335506 5 g
AAB2335514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2335522 Supplier Thermo Scientific Chemicals Supplier No. B2335522
Add to Cart

CAS: 53279-72-4 | C7H3I3O3 | 515.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53279-72-4
Molecular Formula C7H3I3O3
Molecular Weight (g/mol) 515.81
MDL Number MFCD00020305
InChI Key GIAVHGFPMPSIFI-UHFFFAOYSA-N
Synonym htba, 2,4,6-triiodo-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-2,4,6-triiodo, 3-hydroxy-2,4,6-triiodobenzoicacid, pubchem17410, acmc-1aozs, ksc496i6r, 3 hydroxy-2,4,6-triiodobenzoic acid, 3-hydroxy-2,4,6 triiodobenzoic acid
PubChem CID 96627
IUPAC Name 3-hydroxy-2,4,6-triiodobenzoic acid
SMILES OC(=O)C1=C(I)C=C(I)C(O)=C1I

Specifications

Melting Point 211°C to 215°C
Quantity 100 g
Beilstein 2113494
Sensitivity Light sensitive
Formula Weight 515.81
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-2,4,6-triiodobenzoic acid
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 258-457-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.