Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-N-methyl-L-isoleucine, 95%, Thermo Scientific Chemicals

Catalog No. AAH6364006
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6364006 5 g
AAH63640MD 250 mg
AAH6364003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6364006 Supplier Thermo Scientific Chemicals Supplier No. H6364006
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138775-22-1
Molecular Formula C22H25NO4
Molecular Weight (g/mol) 367.45
MDL Number MFCD00153389
InChI Key IQIOLCJHRZWOLS-DZLYFNQKNA-N
Synonym fmoc-n-me-ile-oh, fmoc-n-methyl-l-isoleucine, fmoc-n-me-lle-oh, n-fmoc-n-methyl-l-isoleucine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-isoleucine, 2s,3s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-methylpentanoic acid, fmoc-melle-oh, fmoc-l-meile-oh, ambotzfaa1398, pubchem18966
PubChem CID 15433034
IUPAC Name (2S,3S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-methylpentanoic acid
SMILES CC[C@H](C)[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Beilstein 7548444
Quantity 5 g
Formula Weight 367.44
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Fmoc-N-methyl-L-isoleucine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.