Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-nitrophenylacetic acid, 96%

Catalog No. AAH6481014
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6481014 25 g
AAH6481006 5 g
AAH6481022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6481014 Supplier Thermo Scientific Chemicals Supplier No. H6481014
Add to Cart

CAS: 23876-15-5 | C9H9NO4 | 195.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23876-15-5
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD07782132
InChI Key LWIOFILTAJJDLA-UHFFFAOYSA-N
Synonym 2-methyl-3-nitrophenylacetic acid, 2-methyl-3-nitrophenyl acetic acid, 2-2-methyl-3-nitrophenyl acetic acid, 2-methyl-3-nitro-benzeneacetic acid, 3-nitro-o-tolyl acetic acid, benzeneacetic acid, 2-methyl-3-nitro, 2-methyl-3-nitrophenylaceticacid, 2-methyl-3-nitrobenzeneacetic acid, 2-methyl-3-nitro phenyl acetic acid, pubchem18230
PubChem CID 13530897
SMILES CC1=C(C=CC=C1[N+](=O)[O-])CC(=O)O

Specifications

Melting Point 132°C to 136°C
Quantity 25 g
Beilstein 2581973
Formula Weight 195.17
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Methyl-3-nitrophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.