Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-methyl-3-nitropyridine, 98%

Catalog No. AAH6498214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6498214 25 g
AAH6498206 5 g
AAH6498203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6498214 Supplier Thermo Scientific Chemicals Supplier No. H6498214
Add to Cart

CAS: 374633-31-5 | C6H5BrN2O2 | 217.022 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374633-31-5
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.022
MDL Number MFCD03095212
InChI Key IHHLJCDGOSZFTB-UHFFFAOYSA-N
Synonym 2-bromo-3-nitro-6-methylpyridine, 2-bromo-3-nitro-6-picoline, 2-bromo-6-methyl-3-nitro-pyridine, 2-bromo-3-nitro-6-methyl pyridine, pubchem5954, acmc-209it3, ksc496c9p, 6-bromo-5-nitro-2-picoline, pyridine, 2-bromo-6-methyl-3-nitro, 2-bromo-6-methyl-3-nitropyridinemethyl 6-bromomethyl nicotinate
PubChem CID 24728013
IUPAC Name 2-bromo-6-methyl-3-nitropyridine
SMILES CC1=NC(=C(C=C1)[N+](=O)[O-])Br

Specifications

Color White
Quantity 25 g
Beilstein 138200
Formula Weight 217.02
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-6-methyl-3-nitropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.