Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Serinamide hydrochloride, 95%

Catalog No. AAH6516914
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6516914 25 g
AAH6516906 5 g
AAH6516922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6516914 Supplier Thermo Scientific Chemicals Supplier No. H6516914
Add to Cart

CAS: 65414-74-6 | C3H9ClN2O2 | 140.567 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65414-74-6
Molecular Formula C3H9ClN2O2
Molecular Weight (g/mol) 140.567
MDL Number MFCD00067560
InChI Key VURWDDZIWBGXCK-DKWTVANSSA-N
Synonym l-serinamide hydrochloride, h-ser-nh2.hcl, l-serine amide hydrochloride, s-2-amino-3-hydroxypropanamide hydrochloride, 2s-2-amino-3-hydroxypropanamide hydrochloride, propanamide, 2-amino-3-hydroxy-, monohydrochloride, 2s, s-2-amino-3-hydroxypropionamide hydrochloride, h-ser-nh hcl, h-ser-nh2cl, h-ser-nh2?cl
PubChem CID 16212403
IUPAC Name (2S)-2-amino-3-hydroxypropanamide;hydrochloride
SMILES C(C(C(=O)N)N)O.Cl

Specifications

Melting Point 185°C to 188°C
Beilstein 3911773
Quantity 25 g
Formula Weight 140.57
Percent Purity 95%
Physical Form Powder
Chemical Name or Material L-Serinamide hydrochloride
EINECSNumber 265-753-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.