Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-methylphenol, 97%, Thermo Scientific Chemicals

Catalog No. AAH6625222
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6625222 100 g
AAH6625236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6625222 Supplier Thermo Scientific Chemicals Supplier No. H6625222
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2835-95-2
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00043922
InChI Key DBFYESDCPWWCHN-UHFFFAOYSA-N
Synonym 5-amino-o-cresol, 4-amino-2-hydroxytoluene, 3-hydroxy-4-methylaniline, phenol, 5-amino-2-methyl, 3-amino-6-methylphenol, 6-methyl-3-aminophenol, unii-ubb8xeb10b, 5-amino-2-methyl-phenol, 5-amino-2-methyl phenol, ccris 4582
PubChem CID 17818
IUPAC Name 5-amino-2-methylphenol
SMILES CC1=C(C=C(C=C1)N)O

Specifications

Melting Point 160°C to 162°C
Color Brown
Quantity 100 g
Beilstein 2802317
Formula Weight 123.16
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 5-Amino-2-methylphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-618-6
RTECSNumber SJ6090000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.