Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-L-aspartic acid 1-benzyl ester, 95%

Catalog No. AAH6655806
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6655806 5 g
AAH6655803 1 g
AAH66558MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6655806 Supplier Thermo Scientific Chemicals Supplier No. H6655806
Add to Cart

CAS: 86060-83-5 | C26H23NO6 | 445.471 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86060-83-5
Molecular Formula C26H23NO6
Molecular Weight (g/mol) 445.471
MDL Number MFCD00198201
InChI Key CBZSVHFNEMONDZ-QHCPKHFHSA-N
Synonym fmoc-asp-obzl, fmoc-l-aspartic acid alpha-benzyl ester, s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy-4-oxobutanoic acid, fmoc-l-aspartic acid-1-benzyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-aspartic acid alpha-benzyl ester, 3s-4-benzyloxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-l-asp-obzl, fmoc-aspartic acid-obzl, pubchem14955, fmoc-l-aspartic, a-benzyl ester
PubChem CID 11224591
IUPAC Name (3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-phenylmethoxybutanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Color White
Beilstein 4606552
Quantity 5 g
Formula Weight 445.47
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-L-aspartic acid 1-benzyl ester
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.