Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sarafloxacin hydrochloride hydrate, 98%

Catalog No. AAJ66639MC
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ66639MC 100 mg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66639MC Supplier Thermo Scientific Chemicals Supplier No. J66639MC
Add to Cart

CAS: 91296-87-6 | C20H18ClF2N3O3 | 421.829 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91296-87-6
Molecular Formula C20H18ClF2N3O3
Molecular Weight (g/mol) 421.829
MDL Number MFCD11869789
InChI Key KNWODGJQLCISLC-UHFFFAOYSA-N
Synonym sarafloxacin hydrochloride, sarafloxacin hcl, unii-i36jp4q9df, abbott-56620, i36jp4q9df, sarafloxacin hydrochloride usan, saraflox, dsstox_cid_25939, dsstox_rid_81237
PubChem CID 56207
IUPAC Name 6-fluoro-1-(4-fluorophenyl)-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid;hydrochloride
SMILES C1CN(CCN1)C2=C(C=C3C(=C2)N(C=C(C3=O)C(=O)O)C4=CC=C(C=C4)F)F.Cl

Specifications

Melting Point 275°C
Flash Point >110°C (230°F)
Packaging Plastic bottle
Quantity 100 mg
Beilstein 6495500
Solubility Information Soluble in water
Formula Weight 475.87
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sarafloxacin hydrochloride hydrate
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.