Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ticaracillin disodium salt, 50mg/mL in distilled water, sterile-filtered, Thermo Scientific™

Catalog No. AAJ67144XF
Change view
Click to view available options
Quantity:
1 mL
5 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67144XF 1 mL
AAJ671448EQ 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67144XF Supplier Thermo Scientific Chemicals Supplier No. J67144XF
Add to Cart

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4697-14-7
Molecular Formula C15H14N2Na2O6S2
Molecular Weight (g/mol) 428.38
MDL Number MFCD07787410,MFCD07787410
InChI Key ZBBCUBMBMZNEME-QBGWIPKPSA-L
Synonym ticarcillin disodium, ticillin, ticarcillin disodium usan:usp, n-2-carboxy-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 hept-6-yl-3-thiophenemalonamic acid disodium salt, 3-thiophenemalonamic acid, n-2-carboxy-3,3-dimethyl-7-oxo-4-thia-1-azabicylo 3.2.0 hept-6-yl-, disodium salt, 4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid, 6-carboxy-3-thienylacetyl amino-3,3-dimethyl-7-oxo-, disodium salt, 2s-2alpha,5alpha,6beta s*, disodium 2s,5r,6r-3,3-dimethyl-6-3-oxido-3-oxo-2-thiophen-3-ylpropanoyl amino-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylate
PubChem CID 656842
IUPAC Name disodium (2S,5R,6R)-6-[(2R)-2-carboxylato-2-(thiophen-3-yl)acetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](C([O-])=O)C3=CSC=C3)C(=O)N2[C@H]1C([O-])=O

Specifications

Physical Form Liquid
Quantity 1 mL
Packaging Plastic bottle
Chemical Name or Material Ticaracillin disodium salt
Hazard Category H317-H334
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
RTECSNumber XM9410000
TSCA No
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.