Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cephalexin, 50mg/mL in 1M ammonium hydroxide, sterile-filtered, Thermo Scientific™

Catalog No. AAJ67377AC
Change view
Click to view available options
Quantity:
25 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67377AC 25 mL
AAJ67377AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67377AC Supplier Thermo Scientific Chemicals Supplier No. J67377AC
Add to Cart

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15686-71-2
Molecular Formula C16H20ClN3O5S
Molecular Weight (g/mol) 401.862
MDL Number MFCD00167148
InChI Key YHJDZIQOCSDIQU-OEDJVVDHSA-N
PubChem CID 57515946
IUPAC Name (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydron;chloride;hydrate
SMILES [H+].CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=CC=C3)N)SC1)C(=O)O.O.[Cl-]

Specifications

Type Cephalexin
Physical Form Liquid
Quantity 25 mL
Chemical Name or Material Cephalexin, 50mg/mL in 1M ammonium hydroxide, sterile-filtered
Hazard Category H315-H318
Hazard Statement GHS H Statement
H334-H318-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
EINECSNumber 239-773-6
RTECSNumber XI0350000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.