Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl phenylcyanoacetate, 98%

Catalog No. AAL0392914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0392914 25 g
AAL0392906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0392914 Supplier Thermo Scientific Chemicals Supplier No. L0392914
Add to Cart

CAS: 4553-07-5 | C11H11NO2 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4553-07-5
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00001866
InChI Key SXIRJEDGTAKGKU-UHFFFAOYNA-N
Synonym ethyl phenylcyanoacetate, phenylcyanoacetic acid ethyl ester, ethyl cyanophenylacetate, ethyl cyano phenyl acetate, ethyl alpha-cyanophenylacetate, ethyl alpha-cyano-alpha-phenylacetate, acetic acid, cyanophenyl-, ethyl ester, alpha-cyanobenzeneacetic acid, ethyl ester, benzeneacetic acid, alpha-cyano-, ethyl ester, benzeneacetic acid, .alpha.-cyano-, ethyl ester
PubChem CID 95298
ChEBI CHEBI:51926
IUPAC Name ethyl 2-cyano-2-phenylacetate
SMILES CCOC(=O)C(C#N)C1=CC=CC=C1

Specifications

Density 1.09
Boiling Point 274°C to 275°C
Flash Point >110°C (230°F)
Refractive Index 1.505
Quantity 25 g
Beilstein 611232
Formula Weight 189.21
Percent Purity 98%
Chemical Name or Material Ethyl phenylcyanoacetate
Hazard Category H302+H312+H332-H315-H319
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 224-921-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.