Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nonanol, 98%

Catalog No. AAL0581214
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0581214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0581214 Supplier Thermo Scientific Chemicals Supplier No. L0581214
Add to Cart

CAS: 623-93-8 | C9H20O | 144.258 g/mol

5-Nonanol is used as intermediates in chemical research. It is used as natural substances and extractives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nonanol is used as intermediates in chemical research. It is used as natural substances and extractives.

Solubility
Soluble in water (0.46 mg/ml at 15°C), and chloroform.

Notes
Incompatible ma
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-93-8
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.258
MDL Number MFCD00021944
InChI Key FCBBRODPXVPZAH-UHFFFAOYSA-N
Synonym 5-nonanol, dibutylcarbinol, dibutyl carbinol, 5-nonyl alcohol, nonanol-5, acmc-1bc2t, di-n-butyl carbinol, 5-nonanol 25g, 5-nonanol gc, 5-nonanol, puriss gc
PubChem CID 12202
IUPAC Name nonan-5-ol
SMILES CCCCC(CCCC)O

Specifications

Melting Point 5°C
Density 0.826
Boiling Point 194°C to 196°C
Flash Point 96°C (204°F)
Refractive Index 1.429
Quantity 25 g
Beilstein 1733552
Solubility Information Soluble in water (0.46mg/ml at 15°C),and chloroform.
Formula Weight 144.26
Percent Purity 98%
Chemical Name or Material 5-Nonanol
Show More Show Less
Hazard Category H227-H315-H319
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 210-820-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.