Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myelin Basic Protein Chicken anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB9348MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9348MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9348MI Supplier MilliporeSigma Supplier No. AB9348
Only null left
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Anti-Myelin Basic Protein Antibody detects level of Myelin Basic Protein and has been published and validated for use in immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myelin Basic Protein
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified chicken IgY. Liquid in PBS, pH 7.2 with 0.02% sodium azide.
Gene Symbols MBP; MGC99675; Golli-mbp
Host Species Chicken
Immunogen Synthetic peptide from human/mouse MBP.
Purification Method Affinity Purified
Quantity 500 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001025094.1; NM_001025098.1; NM_001025081.1; NM_001025090.1; NM_001025092.1; NM_001025100.1; NM_001025101.1; NM_002385.2
Target Species Human, Mouse
Content And Storage 2-8°C for 6 months in undiluted aliquotes.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.