Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-oxocyclopentylacetate, 95%

Catalog No. AC123040050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC123040050 5 g Glass bottle
AC123040010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC123040050 Supplier Thermo Scientific Chemicals Supplier No. 123040050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 20826-94-2 | C9H14O3 | 170.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20826-94-2
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00044715
InChI Key PJMKFKUFBDXYEC-SSDOTTSWSA-N
Synonym ethyl 2-oxocyclopentylacetate, ethyl 2-2-oxocyclopentyl acetate, ethyl 2-oxocyclopentaneacetate, ethyl 2-oxocyclopentyl acetate, 2-oxocyclopentaneacetic acid ethyl ester, cyclopentaneacetic acid, 2-oxo-, ethyl ester, pubchem23864, ksc201q9p, ethyl 2-oxo-cyclopentyl acetate, ehtyl 2-2-oxocyclopentyl acetate
PubChem CID 2723663
IUPAC Name ethyl 2-(2-oxocyclopentyl)acetate
SMILES CCOC(=O)C[C@H]1CCCC1=O

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4507 to 1.4527
Quantity 5 g
Formula Weight 170.21
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl 2-oxocyclopentylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.