Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals L(-)-Tryptophan, 99%

Catalog No. AC140590050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Packaging:
Glass Bottle
Plastic Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC140590050 5 g Glass Bottle
AC140590250 25 g Plastic Bottle
AC140591000 100 g Plastic Bottle
AC140595000 500 g Plastic Bottle
AC140590010 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC140590050 Supplier Thermo Scientific Chemicals Supplier No. 140590050
Add to Cart

L(-)-Tryptophan, CAS # 73-22-3, is an essential amino acid necessary for the biosynthesis of proteins. | CAS: 73-22-3 | C11H12N2O2 | 204.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L(-)-Tryptophan is a nonpolar aromatic compound and essential amino acid required for protein synthesis
  • It is a biochemical precursor of niacin, indole alkaloids, and serotonin

Applications

  • L-Tryptophan acts as a natural dietary supplement and has laboratory applications in the research of antidepressants, anxiolytics, and sleep aids
  • It is a precursor of metabolites that are important in the laboratory study of mammalian physiology, including gastrointestinal functions, immunity, metabolism, and the nervous system
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-22-3
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00064340
InChI Key QIVBCDIJIAJPQS-VIFPVBQESA-N
Synonym l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
PubChem CID 6305
ChEBI CHEBI:16828
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point 280.0°C to 285.0°C
Color White to White-Yellow
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (1 g, 105°C)
Assay Percent Range 99%
Packaging Glass Bottle
Specific Rotation − 31.10
Beilstein 22, 546
Merck Index 15, 9977
Solubility Information Solubility in water: 11.4g/L (25°C). Other solubilities: soluble in alkali hydroxides and hot alcohol, insoluble in chloroform
Specific Rotation Condition − 31.10 (20.00°C c=1,H20)
Quantity 5 g
Formula Weight 204.23
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material L(-)-Tryptophan, 99%
Show More Show Less
EINECSNumber 200-795-6
RTECSNumber YN6130000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.