Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Piperidineethanol, 95%

Catalog No. AC157615000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157615000 500 g Glass bottle
AC157610050 5 g Glass bottle
AC157611000 100 g Glass bottle
AC157610025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC157615000 Supplier Thermo Scientific Chemicals Supplier No. 157615000
Add to Cart

CAS: 1484-84-0 | C7H15NO | 129.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1484-84-0
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.2
InChI Key PTHDBHDZSMGHKF-UHFFFAOYSA-N
Synonym 2-piperidineethanol, 2-2-hydroxyethyl piperidine, 2-piperidin-2-yl ethanol, 2-2-piperidyl ethan-1-ol, 2-piperidine ethanol, 2-hydroxyethyl piperidine, 2-piperidine-ethanol, piperidine-2-ethanol, 2-piperidin-2-yl ethan-1-ol
PubChem CID 15144
IUPAC Name 2-piperidin-2-ylethanol
SMILES C1CCNC(C1)CCO

Specifications

Melting Point 38.0°C to 43.0°C
Color White to Yellow
Density 1.0100g/mL
Boiling Point 234.0°C
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 500 g
Beilstein 21,2
Specific Gravity 1.01
Solubility Information Solubility in water: very soluble. Other solubilities: soluble in alcohol
Formula Weight 129.2
Percent Purity 95%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 2-Piperidineethanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 216-059-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.