Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxybenzyl alcohol, 97%

Catalog No. AC173331000
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173331000 100 g Glass bottle
AC173330250 25 g Glass bottle
AC173330010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC173331000 Supplier Thermo Scientific Chemicals Supplier No. 173331000
Add to Cart

CAS: 90-01-7 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-01-7
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00004617
InChI Key CQRYARSYNCAZFO-UHFFFAOYSA-N
Synonym salicyl alcohol, 2-hydroxybenzyl alcohol, saligenin, 2-hydroxymethyl phenol, 2-methylolphenol, o-hydroxybenzyl alcohol, saligenol, benzenemethanol, 2-hydroxy, diathesin, o-methylolphenol
PubChem CID 5146
ChEBI CHEBI:16464
IUPAC Name 2-(hydroxymethyl)phenol
SMILES OCC1=CC=CC=C1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 83.0°C to 87.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula HOC6H4CH2OH
Quantity 100 g
Beilstein 06,891
Fieser 17,305
Merck Index 15,8461
Solubility Information Solubility in water: 67g/L (22 c). Other solubilities: soluble in alcohol,chloroform,ether,propylene,glycol,benzene and fixed oils
Formula Weight 124.14
Percent Purity 97%
Physical Form Crystalline Flakes or Powder
Chemical Name or Material 2-Hydroxybenzyl alcohol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-960-5
RTECSNumber DO6430000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.