Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxytryptamine, 99%

Catalog No. AC187452500
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC187452500 250 mg Glass bottle
AC187450010 1 g Glass bottle
AC187450050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC187452500 Supplier Thermo Scientific Chemicals Supplier No. 187452500
Add to Cart

CAS: 3610-36-4 | C11H14N2O | 190.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3610-36-4
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.24
MDL Number MFCD00005663
InChI Key VOCGEKMEZOPDFP-UHFFFAOYSA-N
PubChem CID 17654
IUPAC Name 2-(6-methoxy-1H-indol-3-yl)ethanamine
SMILES COC1=CC2=C(C=C1)C(=CN2)CCN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 143.0°C to 146.0°C
Assay Percent Range 99%
Packaging Glass bottle
Quantity 250 mg
Synonym 6-methoxytryptamine, 2-6-methoxy-1h-indol-3-yl ethanamine, 3-2-aminoethyl-6-methoxyindole, 2-6-methoxy-1h-indol-3-yl-ethylamine, 1h-indole-3-ethanamine, 6-methoxy, 2-2-aminoethyl-5-methoxyindole, 1h-indole-3-ethanamine,6-methoxy, 2-6-methoxy-1h-indol-3-yl ethan-1-amine, 2-6-methoxyindol-3-yl ethylamine, 6-methoxy tryptamine
Solubility Information Solubility in water: moderately soluble. Other solubilities: soluble in ethanol
Formula Weight 190.24
Percent Purity 99%
Chemical Name or Material 6-Methoxytryptamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 222-778-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.