Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Fuchsin

Catalog No. AC212141000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC212141000 100 g Glass bottle
AC212140250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC212141000 Supplier Thermo Scientific Chemicals Supplier No. 212141000
Add to Cart

CAS: 3248-91-7 | C22H24ClN3 | 365.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3248-91-7
Molecular Formula C22H24ClN3
Molecular Weight (g/mol) 365.91
MDL Number MFCD00012567
InChI Key ORDGVBRMUJCHEO-UHFFFAOYSA-M
Synonym Basic Violet 2, C.I. 42520, Magenta III
PubChem CID 14454445
ChEBI CHEBI:87671
IUPAC Name 4-[(4-amino-3-methylphenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-methylaniline;hydrochloride
SMILES [Cl-].CC1=CC(=CC=C1N)[C+](C1=CC=C(N)C(C)=C1)C1=CC=C(N)C(C)=C1

Specifications

Absorbance 1500 (at lambda min.)
Absorption Ratio 1.1 to 1.4
Infrared Spectrum Authentic
Lambda Max 550 to 554nm (in 50% ethanol)
Formula Weight 365.89
Packaging Glass bottle
Color Green
Quantity 100 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material New Fuchsin
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 221-831-7
RTECSNumber CX9825000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.