Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bicarbonate, 99.5%, for analysis

Catalog No. AC217125000
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC217125000 500 g Plastic bottle
AC217120010 1 kg Plastic bottle
AC217120025 2.5 kg Plastic bottle
AC217120100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC217125000 Supplier Thermo Scientific Chemicals Supplier No. 217125000
Add to Cart

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O

Specifications

Melting Point 270.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 500 g
Assay Percent Range 99.50%
Linear Formula NaHCO3
Fieser 03,260; 05,595; 11,476; 17,180
Merck Index 15, 8719
Solubility Information Solubility in water: 9 g/100mL (20°C). Other solubilities: 16.4 g/100mL water (60°C), insoluble in alcohol
IUPAC Name sodium;hydrogen carbonate
Formula Weight 84.01
Percent Purity 99.5%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Sodium bicarbonate
Show More Show Less
EINECSNumber 205-633-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.