Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenhydramine hydrochloride, 99%

Catalog No. AC350851000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC350851000 100 g Glass bottle
AC350850250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC350851000 Supplier Thermo Scientific Chemicals Supplier No. 350851000
Add to Cart

CAS: 147-24-0 | C17H22ClNO | 291.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-24-0
Molecular Formula C17H22ClNO
Molecular Weight (g/mol) 291.82
MDL Number MFCD00012479
InChI Key PCHPORCSPXIHLZ-UHFFFAOYSA-N
Synonym diphenhydramine hydrochloride, dimedrol, diphenhydramine hcl, carphenamine, bendylate, carphenex, cathejell, denydryl, diphamine, eldadryl
PubChem CID 8980
ChEBI CHEBI:4637
IUPAC Name 2-benzhydryloxy-N,N-dimethylethanamine;hydrochloride
SMILES [H+].[Cl-].CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 168°C to 172°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 3339
Solubility Information Solubility in water: 1000g/L. Other solubilities: 500g/L ethanol
Formula Weight 291.82
Percent Purity 99%
Physical Form Adhering Crystals or Crystalline Powder
Chemical Name or Material Diphenhydramine hydrochloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-687-2
RTECSNumber KR7000000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.