Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2R,4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid methyl ester, 95%, 98% ee, Thermo Scientific Chemicals

Catalog No. AC362970010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC362970010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC362970010 Supplier Thermo Scientific Chemicals Supplier No. 362970010
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 321744-26-7
Molecular Formula C12H21NO5
Molecular Weight (g/mol) 259.30
MDL Number MFCD02259744
InChI Key RNMVWSAJMIKMDY-DTWKUNHWSA-N
PubChem CID 2734504
IUPAC Name 1-O-tert-butyl 2-O-methyl (2R,4S)-4-hydroxypiperidine-1,2-dicarboxylate
SMILES COC(=O)[C@H]1C[C@@H](O)CCN1C(=O)OC(C)(C)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Amber to Colorless
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Synonym 2r,4s-n-boc-4-hydroxypiperidine-2-carboxylic acid methyl ester, 2r,4s-1-tert-butyl 2-methyl 4-hydroxypiperidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2r,4s-4-hydroxypiperidine-1,2-dicarboxylate, 2r,4s-n-boc-4-hydroxypiperidine-2-carboxylic acid, 2r,4s-1-tert-butyl2-methyl4-hydroxypiperidine-1,2-dicarboxylate, n-tert-butoxycarbonyl-cis-4-hydroxy-l-pipecolinic acid methyl ester, 1-o-tert-butyl 2-o-methyl 2r,4s-4-hydroxypiperidine-1,2-dicarboxylate, o1-tert-butyl o2-methyl 2r,4s-4-oxidanylpiperidine-1,2-dicarboxylate, tert-butyl 4s,2r-4-hydroxy-2-methoxycarbonyl piperidinecarboxylate, 2r-4beta-hydroxypiperidine-1,2beta-dicarboxylic acid 1-tert-butyl 2-methyl ester
Formula Weight 259.3
Percent Purity 95%,98% ee
Physical Form Viscous Liquid or Waxy Mass
Chemical Name or Material (2R, 4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid methyl ester, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.