Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™

Catalog No. AC397820050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397820050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC397820050 Supplier Thermo Scientific Chemicals Supplier No. 397820050
Add to Cart

Chemical Identifiers

CAS 126917-10-0
Molecular Formula C8H3ClF4O
Molecular Weight (g/mol) 226.56
MDL Number MFCD00061155
InChI Key OOAHPLWBUUTFMV-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethyl benzoyl chloride, 2-fluoro-4-trifluoromethyl benzoylchloride, benzoyl chloride, 2-fluoro-4-trifluoromethyl, 2-fluoro-4-trifluoromethyl-benzoyl chloride, benzoyl chloride,2-fluoro-4-trifluoromethyl, 2-fluoro-4-trifluoromethyl-1-benzenecarbonyl chloride, alpha,alpha,alpha,2-tetrafluoro-p-toluoyl chloride, acmc-1c5cr, 2-fluoro-4-trifluoromethyl-benzoylchloride, 4-trifluoromethyl-2-fluorobenzoyl chloride
PubChem CID 145580
IUPAC Name 2-fluoro-4-(trifluoromethyl)benzoyl chloride
SMILES C1=CC(=C(C=C1C(F)(F)F)F)C(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.5000g/mL
Boiling Point 188.0°C to 189.0°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4650 to 1.4670 (20°C, 589nm)
Linear Formula FC6H3(CF3)COCl
Quantity 5 g
Specific Gravity 1.5
Formula Weight 226.56
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.