Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) sulfide, 99%, pure

Catalog No. AC453995000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453995000 500 g
AC453991000 100 g
AC453990025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC453995000 Supplier Thermo Scientific Chemicals Supplier No. 453995000
Add to Cart

CAS: 1345-07-9 | Bi2S3 | 514.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1345-07-9
Molecular Formula Bi2S3
Molecular Weight (g/mol) 514.14
MDL Number MFCD00014204
InChI Key YNRGZHRFBQOYPP-UHFFFAOYSA-N
IUPAC Name dibismuth(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Bi+3].[Bi+3]

Specifications

Quantity 500 g
Assay Percent Range 80.1 to 82.5% (Bi) (Complexometry)
Linear Formula Bi2S3
Merck Index 15,1287
Formula Weight 514.16
Percent Purity 99%
Grade Pure
Chemical Name or Material Bismuth(III) sulfide
EINECSNumber 215-716-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.