Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thallium AA Standard, 1000 ppm Tl in 3% HNO3, Ricca Chemical
SDP

Catalog No. ATL1KN500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ATL1KN-500 500 mL Natural Poly Bottle
ATL1KN-100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ATL1KN-500 Supplier Ricca Chemical Company Supplier No. ATL1KN500
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7440-28-0 , 7697-37-2 , 7732-18-5
Molecular Formula Tl
Molecular Weight (g/mol) 204.38
MDL Number MFCD00134063
InChI Key BKVIYDNLLOSFOA-UHFFFAOYSA-N
PubChem CID 5359464
ChEBI CHEBI:30440
IUPAC Name thallium
SMILES [Tl]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 1,000ppm Tl
CAS Min % 93.0, 4.92, 0.1
CAS Max % 97.0, 5.12, 0.1
Equivalence 1 mL = 1 mg Tl
Physical Form Liquid
Color Colorless
Quantity 500 mL
Chemical Name or Material Thallium AA Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.