Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butyric acid

Catalog No. B103500 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B103500 500 mL
B103500100M 100 mL
B1035005ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B103500 Supplier Sigma Aldrich Supplier No. B103500500ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Butyric acid

Specifications

Melting Point -6°C to +3°C (lit.)
CAS 107-92-6
Boiling Point 162°C (lit.)
Molecular Formula C4H8O2
Linear Formula CH3CH2CH2COOH
MDL Number MFCD00002814
Molecular Weight (g/mol) 88.11
Percent Purity ≥99%
Density 0.964 g/mL (at 25°C (literature))
Quantity 500 mL
Refractive Index n20/D 1.398 (literature)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.