Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bromobenzene

Catalog No. B577022L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 L
18 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B577022L 2 L
B57702500ML 500 mL
B57702100ML 100 mL
B5770218L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. B577022L Supplier Sigma Aldrich Supplier No. B577022L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bromobenzene

Specifications

CAS 108-86-1
Melting Point -31°C (lit.)
Density 1.474 g/mL at 25°C, 1.491 g/mL (at 25°C (literature))
Boiling Point 156°C (lit.)
Molecular Formula C6H5Br
Refractive Index n20/D 1.559 (literature)
Linear Formula C6H5Br
MDL Number MFCD00000055
Quantity 2 L
Molecular Weight (g/mol) 157.01
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.